Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitrophthalic anhydride tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Nitroisobenzofuran-1,3-dione

L08020-14 L08020-06 L08020-06 L08020-14
AAAL08020-06EA 67.39 USD InStock
AAAL08020-06 CAAAAL08020-14 AAAL08020-14 CAAAAL08020-06
4-Nitrophthalic anhydride tech. 90%, Technical Grade
4-Nitrophthalic anhydride


Formula: C₈H₃NO₅
MW: 193.12 g/mol
Melting Pt: 116…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00005922
CAS Number: 5466-84-2
EINECS: 226-776-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty