Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-ɑ-D-galactopyranoside monohydrate

Supplier: Thermo Scientific Chemicals
Synonyms: Methyl-ɑ-D-galactoside monohydrate

L08049-03 L08049-06 L08049-03 L08049-06
AAAL08049-03EA 20.01 USD InStock
CAAAAL08049-03 AAAL08049-03 CAAAAL08049-06 AAAL08049-06
Methyl-ɑ-D-galactopyranoside monohydrate
Methyl-alpha-D-galactopyranoside monohydrate


Formula: C₇H₁₄O₆·H₂O
MW: 194.18 g/mol (anhy)
Melting Pt: 105…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064085
CAS Number: 34004-14-3
EINECS: 222-251-7
Order Now
Specification Test Results
Optical rotation +169° (c=1.5 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty