Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-β-D-galactopyranoside

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triolMethyl-β-D-galactoside(2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

L08127-06 L08127-06 L08127-03 L08127-03
AAAL08127-06EA 165 USD InStock
AAAL08127-06 CAAAAL08127-03 AAAL08127-03 CAAAAL08127-06
Methyl-β-D-galactopyranoside
Methyl-beta-D-galactopyranoside


Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Melting Pt: 175…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064357
CAS Number: 1824-94-8
EINECS: 217-361-7
Order Now
Specification Test Results
Optical rotation -16° (c=1.5 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty