Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-5-(trifluoromethyl)pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-5-trifluoromethylpyridine

L08207-14 L08207-14 L08207-22 L08207-22
AAAL08207-14EA 66.23 USD InStock
CAAAAL08207-14 AAAL08207-14 CAAAAL08207-22 AAAL08207-22
2-Chloro-5-(trifluoromethyl)pyridine 97%
2-Chloro-5-(trifluoromethyl)pyridine


Formula: C₆H₃ClF₃N
MW: 181.54 g/mol
Boiling Pt: 152…153 °C
Melting Pt: 29…33 °C
Density: 1.417
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty