Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-3-(trifluoromethyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: α,α,α-Trifluoro-4-nitro-m-cresol5-Hydroxy-2-nitrobenzotrifluoride

L08228-03 L08228-03 L08228-06 L08228-06
AAAL08228-06EA 106.99 USD InStock
CAAAAL08228-06 AAAL08228-06 CAAAAL08228-03 AAAL08228-03
4-Nitro-3-(trifluoromethyl)phenol 97%
4-Nitro-3-(trifluoromethyl)phenol


Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 135…138 °C/0.01 mm
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00014714
CAS Number: 88-30-2
EINECS: 201-818-2
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty