Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Chloro-4-nitrobenzene-1-carbonyl chloride2-Chloro-4-nitrobenzoylchloride

L08246-06 L08246-18 L08246-06 L08246-18
AAAL08246-06EA 48.08 USD InStock
CAAAAL08246-18 CAAAAL08246-06 AAAL08246-06 AAAL08246-18
2-Chloro-4-nitrobenzoyl chloride 98%
2-Chloro-4-nitrobenzoyl chloride


Formula: C₇H₃Cl₂NO₃
MW: 220.01 g/mol
Boiling Pt: 165 °C/20 mm
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00051515
CAS Number: 7073-36-1
EINECS: 230-367-4
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty