Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) predominantly trans ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(tert-butyl)cyclohexanecarboxylic acid

L08248-06 L08248-14 L08248-06 L08248-14
AAAL08248-06EA 60.29 USD InStock
AAAL08248-06 CAAAAL08248-14 CAAAAL08248-06 AAAL08248-14
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) predominantly trans ≥98%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)


Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042622
CAS Number: 5451-55-8
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty