Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,4'-Difluorobiphenyl 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: PFB No. 154,4-Difluoro-1,1-biphenyl

L08361-06 L08361-06
AAAL08361-06EA 71.99 USD InStock
AAAL08361-06 CAAAAL08361-06
4,4'-Difluorobiphenyl 99%
4,4'-Difluorobiphenyl


Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 254…255 °C
Melting Pt: 87…89 °C
Density: 1.361 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000349
CAS Number: 398-23-2
EINECS: 206-910-6
UN: 3152
ADR: "9",II
Merck Index: 13,03173
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty