Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-L-methionine ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-Met-OHBOC-L-MethionineN-Boc-L-methionine

L08366-06 L08366-03 L08366-06 L08366-03
AAAL08366-03EA 22.99 USD InStock
AAAL08366-03 AAAL08366-06 CAAAAL08366-06 CAAAAL08366-03
N-(tert-Butoxycarbonyl)-L-methionine ≥98%
N-(tert-Butoxycarbonyl)-L-methionine


Formula: C₁₀H₁₉NO₄S
MW: 249.33 g/mol
Melting Pt: 48…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065586
CAS Number: 2488-15-5
EINECS: 219-639-3
Order Now
Specification Test Results
Optical rotation -22° (c=1 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty