Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(3,5-Dimethyl-1H-pyrazol-1-yl)-3-oxopropanenitrile

L08369-06 L08369-14 L08369-06 L08369-14
AAAL08369-06EA 92.39 USD InStock
AAAL08369-06 CAAAAL08369-14 CAAAAL08369-06 AAAL08369-14
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole 97%
1-Cyanoacetyl-3,5-dimethyl-1H-pyrazole


Formula: C₈H₉N₃O
MW: 163.18 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00051954
CAS Number: 36140-83-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty