Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-1,3-propanediol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Serinol2-Amino-1,3-dihydroxy propane2-Aminopropane-1,3-diol

L08426-06 L08426-14 L08426-06 L08426-14
AAAL08426-06EA 71.99 USD InStock
AAAL08426-06 CAAAAL08426-06 AAAL08426-14 CAAAAL08426-14
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol


Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 128 °C/0.8 mm
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
UN: 3259
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty