Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Norbornene-2-carboxylic acid predominantly endo isomer 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bicyclo[2.2.1]hept-5-ene-2-carboxylic Acid

L08444-14 L08444-14 L08444-06 L08444-06
AAAL08444-14EA 76.29 USD InStock
CAAAAL08444-14 AAAL08444-14 CAAAAL08444-06 AAAL08444-06
5-Norbornene-2-carboxylic acid predominantly endo isomer 97%
5-Norbornene-2-carboxylic acid


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 136…138 °C/14 mm
Density: 1.129
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00085356
CAS Number: 120-74-1
EINECS: 204-422-8
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty