Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-L-aspartic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-aspartic AcidBoc-Asp-OH

L08498-03 L08498-06 L08498-03 L08498-06
AAAL08498-03EA 27.89 USD InStock
AAAL08498-03 AAAL08498-06 CAAAAL08498-06 CAAAAL08498-03
N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
N-(tert-Butoxycarbonyl)-L-aspartic acid


Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
Melting Pt: 115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037279
CAS Number: 13726-67-5
EINECS: 237-294-7
Order Now
Specification Test Results
Optical rotation -5.5° (c=1 in methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty