Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-tert-Butylacetamide 98%

Supplier: Thermo Scientific Chemicals

L08505-09 L08505-18 L08505-18 L08505-09
AAAL08505-09EA 76.39 USD InStock
AAAL08505-09 AAAL08505-18 CAAAAL08505-09 CAAAAL08505-18
N-tert-Butylacetamide 98%
N-tert-Butylacetamide


Formula: CH₃CONHC(CH₃)₃
MW: 115.18 g/mol
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00038244
CAS Number: 762-84-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty