Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexyltrimethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (1-Hexyl)trimethylammonium bromideHexyltrimethylammonium Bromide

L08594-06 L08594-06 L08594-14 L08594-14
AAAL08594-06EA 43.8 USD InStock
CAAAAL08594-06 AAAL08594-06 CAAAAL08594-14 AAAL08594-14
Hexyltrimethylammonium bromide 98%
Hexyltrimethylammonium bromide


Formula: C₉H₂₂BrN
MW: 224.18 g/mol
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00051872
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty