Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyloxyacetaldehyde dimethyl acetal 97%

Supplier: Thermo Scientific Chemicals

L08651-09 L08651-18 L08651-18 L08651-09
AAAL08651-09EA 77.09 USD InStock
CAAAAL08651-18 CAAAAL08651-09 AAAL08651-09 AAAL08651-18
Benzyloxyacetaldehyde dimethyl acetal 97%
Benzyloxyacetaldehyde dimethyl acetal


Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
Boiling Pt: 80…82 °C/0.5 mm
Density: 1,035
Storage Temperature: Ambient
MDL Number: MFCD00082233
CAS Number: 127657-97-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty