Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Norvaline 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Nva-OHL(+)-Norvaline(S)-2-Aminopentanoic acid(S)-(+)-Norvaline

L08658-06 L08658-03 L08658-03 L08658-06
AAAL08658-03EA 35.19 USD InStock
CAAAAL08658-06 AAAL08658-03 CAAAAL08658-03 AAAL08658-06
L-Norvaline 99%
L-Norvaline


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064421
CAS Number: 6600-40-4
EINECS: 229-543-3
Merck Index: 12,06813
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty