Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Erucic acid tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Synonyms: (Z)-Docos-13-enoic acid

L08685-30 L08685-18 L08685-18 L08685-30
AAAL08685-30EA 131.99 USD InStock
CAAAAL08685-30 AAAL08685-30 AAAL08685-18 CAAAAL08685-18
Erucic acid tech. 90%, Technical Grade
Erucic acid


Formula: C₂₂H₄₂O₂
MW: 338.58 g/mol
Boiling Pt: 265 °C/15 mm
Melting Pt: 29…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
EINECS: 204-011-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty