Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Diphenylpropane 98%

Supplier: Thermo Scientific Chemicals

L08849-09 L08849-09 L08849-04 L08849-04
AAAL08849-09EA 310.99 USD InStock
AAAL08849-09 CAAAAL08849-09 AAAL08849-04 CAAAAL08849-04
1,3-Diphenylpropane 98%
1,3-Diphenylpropane


Formula: (C₆H₅)CH₂CH₂CH₂(C₆H₅)
MW: 196.29 g/mol
Boiling Pt: 158…159 °C/10 mm
Density: 0.980
Storage Temperature: Ambient
MDL Number: MFCD00043574
CAS Number: 1081-75-0
EINECS: 214-101-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty