Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethoxy-4-methylphenol 95%

Supplier: Thermo Scientific Chemicals
Warning

L08898-04 L08898-09 L08898-04 L08898-09
AAAL08898-09EA 290.99 USD InStock
CAAAAL08898-09 AAAL08898-09 AAAL08898-04 CAAAAL08898-04
2-Ethoxy-4-methylphenol 95%
2-Ethoxy-4-methylphenol


Formula: C₉H₁₂O₂
Boiling Pt: 103 °C/3 mm
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00043697
CAS Number: 2563-07-7
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty