Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Benzyl-L-aspartate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: L-Aspartic Acid 4-Benzyl EsterH-Asp(obzl)-OHL-Aspartic Acid beta-Benzyl EsterL-Aspartic acid beta-benzyl esterH-Asp(OBzl)-OHbeta-Benzyl L-Aspartate

L08956-14 L08956-03 L08956-06 L08956-03 L08956-06 L08956-14
AAAL08956-14EA 268.99 USD InStock
CAAAAL08956-03 CAAAAL08956-14 AAAL08956-14 AAAL08956-03 CAAAAL08956-06 AAAL08956-06
4-Benzyl-L-aspartate 98%
4-Benzyl-L-aspartate


Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: ca 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037208
CAS Number: 2177-63-1
EINECS: 218-541-8
Order Now
Specification Test Results
Optical rotation +27.5° (c=1 in 1N HCl)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty