Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,3,5-Tetramethylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N,N-Dimethyl-3,5-xylidine3,5,N,N-Tetramethylaniline3,5-Dimethyl-N,N-dimethylaniline

L09266-06 L09266-06 L09266-14 L09266-14
AAAL09266-14EA 443.99 USD InStock
CAAAAL09266-06 AAAL09266-14 CAAAAL09266-14 AAAL09266-06
N,N,3,5-Tetramethylaniline 98%
N,N,3,5-Tetramethylaniline


Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 226…228 °C
Density: 0.913
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008307
CAS Number: 4913-13-7
EINECS: 225-539-0
UN: 2810
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty