Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Pentynoic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

L09282-06 L09282-06 L09282-03 L09282-03
AAAL09282-03EA 63.49 USD InStock
CAAAAL09282-06 AAAL09282-03 AAAL09282-06 CAAAAL09282-03
4-Pentynoic acid 95%
4-Pentynoic acid


Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 109…110 °C/30 mm
Melting Pt: 51…58 °C
Flash Pt: 75 °C(167 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 2923
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty