Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: o-Fluorobenzophenone

L09316-14 L09316-06 L09316-06 L09316-14
AAAL09316-14EA 122.99 USD InStock
CAAAAL09316-14 CAAAAL09316-06 AAAL09316-14 AAAL09316-06
2-Fluorobenzophenone 98+%
2-Fluorobenzophenone


Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 149…150 °C/16 mm
Density: 1,186
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000318
CAS Number: 342-24-5
EINECS: 206-440-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty