Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-nitrobenzamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L09380-09 L09380-09 L09380-04 L09380-04
AAAL09380-04EA 42.49 USD InStock
CAAAAL09380-04 AAAL09380-04 CAAAAL09380-09 AAAL09380-09
5-Chloro-2-nitrobenzamide 98+%
5-Chloro-2-nitrobenzamide


Formula: C₇H₅ClN₂O₃
Melting Pt: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00017509
CAS Number: 40763-96-0
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty