Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropylacetic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L09416-03 L09416-06 L09416-06 L09416-03
AAAL09416-06EA 284.99 USD InStock
AAAL09416-06 CAAAAL09416-06 AAAL09416-03 CAAAAL09416-03
Cyclopropylacetic acid 97%
Cyclopropylacetic acid


Formula: C₅H₈O₂
Boiling Pt: 189…191 °C
Density: 1.076
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041544
CAS Number: 5239-82-7
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty