Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Pyrrolidinone-5-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DL-2-Pyrrolidone-5-carboxylic acidDL-Pyroglutamic acid5-Oxopyrrolidine-2-carboxylic acid

L09424-30 L09424-30 L09424-18 L09424-18
AAAL09424-18EA 30.32 USD InStock
AAAL09424-18 CAAAAL09424-18 CAAAAL09424-30 AAAL09424-30
(±)-2-Pyrrolidinone-5-carboxylic acid 99%
(±)-2-Pyrrolidinone-5-carboxylic acid


Formula: C₅H₇NO₃
MW: 129.12 g/mol
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty