Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Propyl methacrylate 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 200 ppm

L09493-14 L09493-06 L09493-06 L09493-14
AAAL09493-14EA 162.99 USD InStock
CAAAAL09493-06 CAAAAL09493-14 AAAL09493-14 AAAL09493-06
n-Propyl methacrylate 95% stabilized
n-Propyl methacrylate


Formula: C₇H₁₂O₂
Boiling Pt: 140…141 °C
Density: 0.902
Flash Pt: 30 °C(86 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048690
CAS Number: 2210-28-8
EINECS: 218-639-0
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty