Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloromethyl chloroformate 97%

Supplier: Thermo Scientific Chemicals
Danger

L09527-09 L09527-18 L09527-09 L09527-18
AAAL09527-18EA 344.99 USD InStock
CAAAAL09527-09 CAAAAL09527-18 AAAL09527-18 AAAL09527-09
Chloromethyl chloroformate 97%
Chloromethyl chloroformate


Formula: C₂H₂Cl₂O₂
Boiling Pt: 107…108 °C
Melting Pt: < –20 °C
Density: 1.465
Flash Pt: 95 °C(203 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00077688
CAS Number: 22128-62-7
EINECS: 244-793-3
UN: 2745
ADR: 6.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty