Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans- mixture 95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,3,4,5-Tetramethylcyclopent-2-enone

L09617-14 L09617-14 L09617-06 L09617-06
AAAL09617-14EA 266.99 USD InStock
CAAAAL09617-14 AAAL09617-14 CAAAAL09617-06 AAAL09617-06
2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans- mixture 95%
2,3,4,5-Tetramethyl-2-cyclopentenone


Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 99…100 °C/30 mm
Density: 0.917
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty