Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-(trifluoromethyl)crotonate mixture E/Z 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Trifluoromethyl)crotonic acid ethyl ester

L09688-04 L09688-09 L09688-09 L09688-04
AAAL09688-09EA 654.99 USD InStock
CAAAAL09688-09 AAAL09688-09 CAAAAL09688-04 AAAL09688-04
Ethyl-3-(trifluoromethyl)crotonate mixture E/Z 96%
Ethyl-3-(trifluoromethyl)crotonate


Formula: C₇H₉F₃O₂
MW: 182.14
Boiling Pt: 129…130 °C
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040846
CAS Number: 24490-03-7
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty