Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Phenylalaninol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D-PenylalaninolD(+)-2-Amino-3-phenyl-1-propanol

L09697-03 L09697-03
AAAL09697-03EA 46.89 USD InStock
AAAL09697-03 CAAAAL09697-03
D-Phenylalaninol 98%
D-Phenylalaninol


Formula: C₉H₁₃NO
MW: 151.2 g/mol
Melting Pt: 89…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00064298
CAS Number: 5267-64-1
EINECS: 226-086-1
UN: 3259
ADR: "8",III
Order Now
Specification Test Results
Optical rotation +23° (c=5 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty