Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Phenylene diisothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: BitoscanatePDITCp-DiisothiocyanatobenzenePhenyl 1,4-diisothiocyanatep-Phenylene diisothiocyanate1,4-Diisothiocyanatobenzene

L09756-03 L09756-06 L09756-03 L09756-06 L09756-14 L09756-14
AAAL09756-03EA 40.29 USD InStock
CAAAAL09756-06 AAAL09756-03 AAAL09756-14 AAAL09756-06 CAAAAL09756-03 CAAAAL09756-14
1,4-Phenylene diisothiocyanate 98%
1,4-Phenylene diisothiocyanate


Formula: C₈H₄N₂S₂
MW: 192.27 g/mol
Melting Pt: 128…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004811
CAS Number: 4044-65-9
EINECS: 223-741-3
UN: 2811
ADR: "6.1",III
Merck Index: 13,01304
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty