Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Cyano-2-methylpyridine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 6-Methylnicotinonitrile5-Cyano-2-picoline

L09875-06 L09875-06 L09875-03 L09875-03
AAAL09875-03EA 58.19 USD InStock
CAAAAL09875-06 CAAAAL09875-03 AAAL09875-03 AAAL09875-06
5-Cyano-2-methylpyridine 99%
5-Cyano-2-methylpyridine


Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 216…217 °C
Melting Pt: 83…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00038042
CAS Number: 3222-48-8
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty