Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Methylthio)phenol 97+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxythioanisole

L10128-14 L10128-06 L10128-03 L10128-03 L10128-14 L10128-06
AAAL10128-06EA 98.59 USD InStock
CAAAAL10128-03 CAAAAL10128-14 CAAAAL10128-06 AAAL10128-06 AAAL10128-14 AAAL10128-03
2-(Methylthio)phenol 97+%
2-(Methylthio)phenol


Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 104…106 °C/22 mm
Density: 1,16
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002211
CAS Number: 1073-29-6
EINECS: 214-027-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty