Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-DL-alanine ≥95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Acetyl-DL-2-aminopropionic AcidAc-DL-Ala-OH

L10329-06 L10329-14 L10329-14 L10329-06
AAAL10329-06EA 25.79 USD InStock
CAAAAL10329-14 AAAL10329-06 AAAL10329-14 CAAAAL10329-06
N-Acetyl-DL-alanine ≥95%
N-Acetyl-DL-alanine


Formula: C₅H₉NO₃
MW: 131.13 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037238
CAS Number: 1115-69-1
EINECS: 214-229-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty