Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2,3,3,3-Pentafluoropropyl acrylate 97%

Supplier: Thermo Scientific Chemicals
Warning

L10343-03 L10343-06 L10343-03 L10343-06
AAAL10343-06EA 58.79 USD InStock
CAAAAL10343-03 AAAL10343-06 CAAAAL10343-06 AAAL10343-03
2,2,3,3,3-Pentafluoropropyl acrylate 97%
2,2,3,3,3-Pentafluoropropyl acrylate


Formula: C₆H₅F₅O₂
Boiling Pt: 49…50 °C/100 mm
Density: 1.320
Flash Pt: 23 °C(73 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00039257
CAS Number: 356-86-5
EINECS: 206-608-4
UN: 3272
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty