Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-piperidinecarboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethyl pipecolinate

L10532-03 L10532-14 L10532-06 L10532-03 L10532-14 L10532-06
AAAL10532-06EA 85.99 USD InStock
AAAL10532-06 AAAL10532-03 AAAL10532-14 CAAAAL10532-06 CAAAAL10532-03 CAAAAL10532-14
Ethyl-2-piperidinecarboxylate 98+%
Ethyl 2-piperidinecarboxylate


Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 216…217 °C
Density: 1.006
Flash Pt: 46 °C(114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005980
CAS Number: 15862-72-3
EINECS: 239-990-6
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty