Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-O-Isopropylidene-ɑ-D-glucofuranose

Supplier: Thermo Scientific Chemicals
Synonyms: (R)-1-((3aR,5R,6S,6aR)-6-Hydroxy-2,2-dimethyltetrahydrofuro[2,3-d][1,3]dioxol-5-yl)ethane-1,2-diol

L10688-14 L10688-06 L10688-06 L10688-14
AAAL10688-14EA 111.99 USD InStock
AAAL10688-14 CAAAAL10688-06 AAAL10688-06 CAAAAL10688-14
1,2-O-Isopropylidene-ɑ-D-glucofuranose
1,2-O-Isopropylidene-alpha-D-glucofuranose


Formula: C₉H₁₆O₆
MW: 220.22 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063244
CAS Number: 18549-40-1
EINECS: 242-420-9
Order Now
Specification Test Results
Optical rotation -13° (c=1 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty