Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Indanylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(2,3-Dihydro-1H-inden-2-yl)acetic acid

L10700-06 L10700-03 L10700-03 L10700-06
AAAL10700-06EA 433.99 USD InStock
CAAAAL10700-06 AAAL10700-06 CAAAAL10700-03 AAAL10700-03
2-Indanylacetic acid 99%
2-Indanylacetic acid


Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00082653
CAS Number: 37868-26-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty