Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-iodoethane 96% stabilized, liquid

Supplier: Thermo Scientific Chemicals
Warning

Stabilization: Stabilised with copper

L10742-03 L10742-14 L10742-14 L10742-03 L10742-06 L10742-06
AAAL10742-14EA 542.99 USD InStock
CAAAAL10742-06 AAAL10742-14 AAAL10742-03 CAAAAL10742-03 CAAAAL10742-14 AAAL10742-06
1-Chloro-2-iodoethane 96% stabilized, liquid
1-Chloro-2-iodoethane


Formula: C₂H₄ClI
MW: 190.41 g/mol
Boiling Pt: 61…62 °C/30 mm
Melting Pt: –15.6 °C
Density: 2.088
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041453
CAS Number: 624-70-4
EINECS: 677-827-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty