Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propiolaldehyde diethyl acetal 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3,3-Diethoxy-1-propynePropioaldehyde diethyl acetal3,3-Diethoxypropyne3,3-Diethoxyprop-1-ynePropargylaldehyde diethyl acetalPropynal diethyl acetal

L10767-06 L10767-03 L10767-06 L10767-14 L10767-03 L10767-14
AAAL10767-06EA 155.99 USD InStock
AAAL10767-06 CAAAAL10767-14 CAAAAL10767-03 AAAL10767-03 CAAAAL10767-06 AAAL10767-14
Propiolaldehyde diethyl acetal 97%
Propiolaldehyde diethyl acetal


Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 138…139 °C
Density: 0.894
Flash Pt: 32 °C(89 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009237
CAS Number: 10160-87-9
EINECS: 233-430-4
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty