Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromophenyl isocyanate 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Bromo-4-isocyanatobenzene

L10834-06 L10834-14 L10834-14 L10834-06
AAAL10834-06EA 48.09 USD InStock
CAAAAL10834-14 AAAL10834-06 CAAAAL10834-06 AAAL10834-14
4-Bromophenyl isocyanate 99%
4-Bromophenyl isocyanate


Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 157…158 °C/14 mm
Melting Pt: 39…42 °C
Flash Pt: 109 °C(228 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002022
CAS Number: 2493-02-9
EINECS: 219-662-9
UN: 3335
ADR: 6.1,II
Merck Index: 13,01415
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty