Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-methylbenzonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

L11048-03 L11048-03 L11048-06 L11048-06
AAAL11048-06EA 112.99 USD InStock
CAAAAL11048-06 AAAL11048-06 CAAAAL11048-03 AAAL11048-03
3-Chloro-2-methylbenzonitrile 97%
3-Chloro-2-methylbenzonitrile


Formula: C₈H₆ClN
MW: 151.60 g/mol
Boiling Pt: 120 °C/22 mm
Melting Pt: 20…23 °C
Storage Temperature: Ambient
MDL Number: MFCD00045598
CAS Number: 54454-12-5
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty