Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diallyl disulfide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Allyl disulphideDiallyldisulfideDiallyldisulphideAllyl disulfide

L11106-22 L11106-14 L11106-22 L11106-14
AAAL11106-22EA 154.99 USD InStock
CAAAAL11106-22 AAAL11106-22 CAAAAL11106-14 AAAL11106-14
Diallyl disulfide tech. 80% remainder mainly diallyl sulfide and diallyl trisulfide, Technical Grade
Diallyl disulphide


Formula: C₆H₁₀S₂
MW: 146.28 g/mol
Boiling Pt: 180…190 °C
Density: 1.008
Flash Pt: 51 °C(123 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008656
CAS Number: 2179-57-9
EINECS: 218-548-6
UN: 1992
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty