Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorothiobenzamide 97%

Supplier: Thermo Scientific Chemicals
Warning

L11180-14 L11180-14 L11180-06 L11180-06
AAAL11180-06EA 90.99 USD InStock
AAAL11180-06 CAAAAL11180-06 AAAL11180-14 CAAAAL11180-14
2-Chlorothiobenzamide 97%
2-Chlorothiobenzamide


Formula: C₇H₆ClNS
MW: 171.65 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00040955
CAS Number: 15717-17-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty