Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

γ-Cyclodextrine hydrate

Supplier: Thermo Scientific Chemicals
Synonyms: γ-Cyclodextrin xhydrateγ-Cyclodextrin hydrate

L11271-ME L11271-ME L11271-MC L11271-MC
AAAL11271-MEEA 136.99 USD InStock
AAAL11271-ME CAAAAL11271-ME AAAL11271-MC CAAAAL11271-MC
γ-Cyclodextrine hydrate
γ-Cyclodextrine hydrate


Formula: C₄₈H₈₀O₄₀·H₂O
MW: 1297.15 g/mol (anhy)
Melting Pt: ca 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149574
CAS Number: 91464-90-3
EINECS: 241-482-4
Order Now
Specification Test Results
Optical rotation +177° (c=1 in water) on dry basis

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty