Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethynylpyridine 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Stabilization: Stabilised with hydroquinone (HQ) 0.01%

L11274-14 L11274-14 L11274-06 L11274-06 L11274-03 L11274-03
AAAL11274-06EA 162.99 USD InStock
AAAL11274-06 AAAL11274-14 AAAL11274-03 CAAAAL11274-14 CAAAAL11274-03 CAAAAL11274-06
2-Ethynylpyridine 98% stabilized
2-Ethynylpyridine


Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 77…78 °C/14 mm
Density: 1.020
Flash Pt: 76 °C(168 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00041598
CAS Number: 1945-84-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty