Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Thio-β-D-glucose tetraacetate ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-mercaptotetrahydro-2H-pyran-3,4,5-triyl triacetate

L11404-MD L11404-03 L11404-MD L11404-03
AAAL11404-MDEA 36.69 USD InStock
AAAL11404-MD CAAAAL11404-MD AAAL11404-03 CAAAAL11404-03
1-Thio-β-D-glucose tetraacetate ≥98%
1-Thio-beta-D-glucose tetraacetate


Formula: C₁₄H₂₀O₉S
MW: 364.37 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063271
CAS Number: 19879-84-6
EINECS: 243-392-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty