Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chlorocinnamamide 97%

Supplier: Thermo Scientific Chemicals

L11406-03 L11406-03 L11406-06 L11406-06
AAAL11406-06EA 239.99 USD InStock
AAAL11406-06 AAAL11406-03 CAAAAL11406-03 CAAAAL11406-06
4-Chlorocinnamamide 97%
4-Chlorocinnamamide


Formula: C₉H₈ClNO
MW: 181.62 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00017147
CAS Number: 18166-64-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty